Learn More
Thermo Scientific™ Infinity Glucose Hexokinase 2 x 125mL
Description
Determine quantity of glucose in human serum, plasma or urine in vitro with Thermo Scientific™ Glucose Hexokinase Reagents.
Diabetes mellitus is a group of metabolic disorders of carbohydrate metabolism in which glucose is underutilized, producing hyperglycemia. Measurement of blood glucose levels plays a key role in the diagnosis and management of the disease. Increased glucose levels may also be observed with hyperactivity of the thyroid, pituitary, or adrenal glands. Decreased levels are present in cases of insulin overdose, insulin secreting tumors, myxedema, hypopituitarism, hypoadrenalism, and conditions interfering with glucose absorption.
- Up to 18 months of stability from the date of manufacture
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Content And Storage | 2°C to 8°C |
| Description | Infinity Glucose Hexokinase |
| CE Marker | Yes |
| Linearity | 0 to 810mg/dL |
| Methods Compatibility | Endpoint |
| Quantity | 2 x 125 mL |
| Stability | Stable Liquid |
| State at Room Temperature | Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.