Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Glucose Oxidase Monoclonal Antibody (GO14-6.6.1.2), Invitrogen™

Catalog No. PIMA546001
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546001 1 mg
PIMA546022 100 μg
01-671-558 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. PIMA546001 Supplier Invitrogen™ Supplier No. MA546001
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-10605 detects Glucose Oxidase from Aspergillus niger samples. MA1-10605 has been successfully used in ELISA applications. The MA1-10605 immunogen is purified glucose oxidase from Aspergillus niger.

Glucose Oxidase Monoclonal antibody specifically detects Glucose Oxidase in Fungi samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glucose Oxidase
Applications ELISA
Classification Monoclonal
Clone GO14-6.6.1.2
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias aspergillus niger glucose oxidase; GOx
Host Species Mouse
Immunogen Purified glucose oxidase from Aspergillus niger.
Purification Method Protein A/G
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Fungi
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.