Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Glucose Solution, 0.1%
SCI_ED

Quantity 50 mL
Packaging 0.10%

 View more versions of this product

Catalog No. S25336


Description

Description

Specifications

Chemical Identifiers

50-99-7, 7732-18-5
180.156
2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
CHEBI:42758
C(C(C(C(C(C=O)O)O)O)O)O
C6H12O6
GZCGUPFRVQAUEE-SLPGGIOYSA-N
107526
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal

Specifications

50-99-7, 7732-18-5
C6H12O6
GZCGUPFRVQAUEE-SLPGGIOYSA-N
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
107526
0.10%
Glucose Solution, 0.1%
0.1% w/v
2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
C(C(C(C(C(C=O)O)O)O)O)O
180.156
CHEBI:42758
50 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 2 years

SDS Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.