Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glucose Standard, 10,000 ppm C6H12O6 in Aqueous Benzoic Acid Solution, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
Specifications
Specifications
Packaging | Natural Poly Bottle |
Odor | Odorless |
Assay Percent Range | 10, 000 ± 50ppm |
Physical Form | Liquid |
Color | Colorless |
Boiling Point | 100°C |
Solubility | Infinitely soluble in water |
Type | Standard |
Solvent or Matrix | Water |
Quantity | 500 mL |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction