Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucose Transporter 8 Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502207213
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-7213 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-7213 Supplier Fabgennix Inc Supplier No. GLUT8801AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Glucose Transporter 8 Polyclonal antibody specifically detects Glucose Transporter 8 in Human, Mouse, Rat samples. It is validated for ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen Glucose Transporter 8
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene SLC2A8
Gene Accession No. Q9JIF3, Q9JJZ1, Q9NY64
Gene Alias D2Ertd44e; Glucose transporter type 8; glucose transporter type X1; glucose transporter X1; Glut8; GLUT-8; GLUTX1; intestinal GLUT8 transporter; SLC2A8; solute carrier family 2 (facilitated glucose transporter), member 8; solute carrier family 2 member 8; solute carrier family 2, (facilitated glucose transporter) member 8; solute carrier family 2, (facilitated glucose transporter), member 8; solute carrier family 2, facilitated glucose transporter member 8
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 400-500 on GLUT8 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29988, 56017, 85256
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.