Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucose Transporter GLUT6 Antibody, Novus Biologicals™
SDP

Catalog No. NBP159891 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP159891 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP159891 Supplier Novus Biologicals Supplier No. NBP159891
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Glucose Transporter GLUT6 Polyclonal specifically detects Glucose Transporter GLUT6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Glucose Transporter GLUT6
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q9UGQ3
Gene Alias Glucose transporter type 6, Glucose transporter type 9, GLUT6, GLUT-9, GLUT9GLUT-6, HSA011372, solute carrier family 2 (facilitated glucose transporter), member 6, solute carrier family 2, facilitated glucose transporter member 6
Gene Symbols SLC2A6
Host Species Rabbit
Immunogen Synthetic peptides corresponding to SLC2A6(solute carrier family 2 (facilitated glucose transporter), member 6) The peptide sequence was selected from the C terminal of SLC2A6. Peptide sequence AANLTLGLYIHFGPRPLSPNSTAGLESESWGDLAQPLAAPAGYLTLVPLL The peptide sequence for this immunogen was taken from within the described region.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11182
Test Specificity Expected identity based on immunogen sequence: Human: 100%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.