Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Glucose Uptake-Glo Assay, 50ml

Catalog No. prj1343 Shop All Promega Corporation Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRJ1343 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRJ1343 Supplier Promega Supplier No. J1343
Only null left
Add to Cart
Add to Cart
Request a Quote

The Glucose Uptake-Glo Assay is a non-radioactive, plate-based, homogeneous bioluminescent method for measuring glucose uptake in mammalian cells based on the detection of 2-deoxyglucose-6-phosphate (2DG6P).

  • A Simple, Non−Radioactive Assay to Measure Glucose Uptake Inside Cells
  • Homogeneous protocol
  • Achieve sensitivity with broad linearity
  • Reliable and reproducible results

Specifications

Content And Storage LESS THAN -65°C
Quantity 50 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.