Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Glucose Uptake-Glo Assay, 5ml

Catalog No. PRJ1341 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRJ1341 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRJ1341 Supplier Promega Supplier No. J1341
May include imposed supplier surcharges.
Add to Cart
Add to Cart

The Glucose Uptake-Glo Assay is a non-radioactive, plate-based, homogeneous bioluminescent method for measuring glucose uptake in mammalian cells based on the detection of 2-deoxyglucose-6-phosphate (2DG6P).

  • A Simple, Non−Radioactive Assay to Measure Glucose Uptake Inside Cells
  • Homogeneous protocol
  • Achieve sensitivity with broad linearity
  • Reliable and reproducible results

Specifications

Content And Storage LESS THAN -65°C
Quantity 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.