Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GluD1/GluD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143863
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143863 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143863 Supplier Invitrogen™ Supplier No. PA5143863
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human HepG2 whole cell, human HL-60 whole cell, human THP-1 whole cell, human CACO-2 whole cell, rat testis tissue, rat C6 whole cell, mouse testis tissue, mouse C2C12 whole cell. IHC: human breast cancer tissue, human endometrial cancer tissue, human lung cancer tissue, human ovarian cancer tissue, human rectal cancer tissue, human renal carcinoma tissue, human gallbladder carcinoma tissue, human gastric carcinoma tissue, human liver cancer tissue, human lymphoma tissue, rat liver tissue. ICC/IF: T47D cell. Flow: SiHa cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Glutamate dehydrogenase (EC 1.4.1.3) catalyzes the reversible oxidative deamination of glutamate to alpha-ketoglutarate using NAD and/or NADP as cofactors. See also GLUD1 (MIM 138130).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GluD1/GluD2
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene GLUD1
Gene Accession No. P00367, P10860, P26443, P49448
Gene Alias Ac2-281; AI118167; epididymis tissue sperm binding protein Li 18mP; GDH; GDH 1; GDH 2; GDH1; GDH2; Gdh-X; Glud; Glud1; GLUD2; Gludeha; Gludl; GLUDP1; glutamate dehydrogenase (NAD(P)+); glutamate dehydrogenase 1; glutamate dehydrogenase 1, mitochondrial; glutamate dehydrogenase 2; glutamate dehydrogenase 2, mitochondrial; glutamate dehydrogenase pseudogene 1; memory-related gene 2 protein; MRG-2; testicular secretory protein Li 14
Gene Symbols GLUD1, GLUD2
Host Species Rabbit
Immunogen E.coli-derived human GLUD1/2 recombinant protein (Position: S54-A553).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14661, 24399, 2746, 2747
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.