Learn More
Invitrogen™ GluD1/GluD2 Recombinant Rabbit Monoclonal Antibody (HL2124)

Description
GluD1/GluD2 Recombinant Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | GluD1/GluD2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | HL2124 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | GLUD1 |
| Gene Accession No. | P00367, P10860, P26443, P49448 |
| Gene Alias | Ac2-281; AI118167; epididymis tissue sperm binding protein Li 18mP; GDH; GDH 1; GDH 2; GDH1; GDH2; Gdh-X; Glud; Glud1; GLUD2; Gludeha; Gludl; GLUDP1; glutamate dehydrogenase (NAD(P)+); glutamate dehydrogenase 1; glutamate dehydrogenase 1, mitochondrial; glutamate dehydrogenase 2; glutamate dehydrogenase 2, mitochondrial; glutamate dehydrogenase pseudogene 1; memory-related gene 2 protein; MRG-2; testicular secretory protein Li 14 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.