Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GluD1/GluD2 Recombinant Rabbit Monoclonal Antibody (HL2125)
GREENER_CHOICE

Catalog No. FEMA550390
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550390 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550390 Supplier Invitrogen™ Supplier No. MA550390
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

GluD1/GluD2 Recombinant Monoclonal Antibody for Western Blot, IHC (P)

Glutamate dehydrogenase (EC 1.4.1.3) catalyzes the reversible oxidative deamination of glutamate to alpha-ketoglutarate using NAD and/or NADP as cofactors. See also GLUD1 (MIM 138130).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GluD1/GluD2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone HL2125
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene GLUD1
Gene Accession No. P00367, P10860, P26443, P49448
Gene Alias Ac2-281; AI118167; epididymis tissue sperm binding protein Li 18mP; GDH; GDH 1; GDH 2; GDH1; GDH2; Gdh-X; Glud; Glud1; GLUD2; Gludeha; Gludl; GLUDP1; glutamate dehydrogenase (NAD(P)+); glutamate dehydrogenase 1; glutamate dehydrogenase 1, mitochondrial; glutamate dehydrogenase 2; glutamate dehydrogenase 2, mitochondrial; glutamate dehydrogenase pseudogene 1; memory-related gene 2 protein; MRG-2; testicular secretory protein Li 14
Gene Symbols GLUD1, GLUD2
Host Species Rabbit
Immunogen Recombinant fragmemt of human GLUD1+GLUD2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14661, 24399, 2746, 2747
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.