Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GluD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576487
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576487 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576487 Supplier Invitrogen™ Supplier No. PA576487
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

Glutamate dehydrogenase, GluD1, is a mitochondrial matrix enzyme that catalyzes the oxidative deamination of glutamate to alpha-ketoglutarate and ammonia. This enzyme has an important role in regulating amino acid-induced insulin secretion. It is allosterically activated by ADP and inhibited by GTP and ATP. Activating mutations in this gene are a common cause of congenital hyperinsulinism. Alternative splicing of this gene results in multiple transcript variants. The related glutamate dehydrogenase 2 gene on the human X-chromosome originated from this gene via retrotransposition and encodes a soluble form of glutamate dehydrogenase. Related pseudogenes have been identified on chromosomes 10, 18 and X.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GluD1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene GLUD1
Gene Accession No. P00367, P10860, P26443
Gene Alias Ac2-281; AI118167; brain glutamate dehydrogenase; epididymis tissue sperm binding protein Li 18mP; GDH; GDH 1; GDH 2; Gdh1; GDH2; Gdh-X; Glud; GLUD1; GLUD1 protein; GLUD2; Gludeha; Gludl; GLUDP1; gluR delta-1 subunit; GluRdelta1; glutamate dehydrogenase; glutamate dehydrogenase (NAD(P)+); glutamate dehydrogenase 1; glutamate dehydrogenase 1, mitochondrial; glutamate dehydrogenase 2; glutamate dehydrogenase 2, mitochondrial; glutamate dehydrogenase pseudogene 1; glutamate ionotropic receptor delta type subunit 1; glutamate receptor delta-1 subunit; glutamate receptor ionotropic, delta-1; glutamate receptor, ionotropic, delta 1; Grid1; KIAA1220; Membrane protein 50; memory-related gene 2 protein; MP50; MRG-2; testicular secretory protein Li 14
Gene Symbols GLUD1
Host Species Rabbit
Immunogen Recombinant fusion protein of human GLUD1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14661, 24399, 2746
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.