Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GluD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA593088
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA593088 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA593088 Supplier Invitrogen™ Supplier No. PA593088
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YLHRFGAKCI AVGESDGSIW NPDGIDPKEL EDFKLQHGSI LGFPKAKPYE GSILEVDCDI LIPAATEKQL TKSNAPRVKA KIIAEGANGP TTPEADKIFL ERNILVIPDL YLNAGGVTVS YFEWLKNLNH VSYGRLTFKY ERDSNYHLLL SVQESLERKF GKHGGTIPIV PTAEFQDSIS GASEKDIVHS ALAYTMERSA RQIMHTAMKY NLGLDLRTAA YVNAIEKVFK VYSEAGVTFT; Positive Samples: A-549, MCF7, THP-1, A-431, Mouse brain, Mouse eye, Rat testis; Cellular Location: Mitochondrion matrix.

Glutamate dehydrogenase 2, Glud2, is localized to the mitochondrion and acts as a homohexamer to recycle glutamate during neurotransmission. The enzyme catalyzes the reversible oxidative deamination of glutamate to alpha-ketoglutarate. Diseases associated with GLUD2 include Parkinson Disease, Late-Onset and Hyperinsulinemic Hypoglycemia, Familial, 6. Among its related pathways are GABA shunt and Carbon metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GluD2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.77 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GLUD2
Gene Accession No. P49448
Gene Alias B230104L07Rik; cpr; creeper; GDH 2; GDH2; GluD2; GLUDP1; GluR delta2; gluR delta-2 subunit; GLURD2; GluRdelta2; glutamate dehydrogenase 2; glutamate dehydrogenase 2, mitochondrial; glutamate dehydrogenase pseudogene 1; glutamate ionotropic receptor delta type subunit 2; glutamate receptor delta-2 subunit; glutamate receptor ionotropic, delta-2; glutamate receptor, ionotropic, delta 2; Grid2; ho; hotfoot; Lc; Lc>J>; lurcher; minisatellite 10ac detected by probe MMS10; MMS10-AC; Ms10ac; nmf408; SCAR18; testicular secretory protein Li 14; tpr
Gene Symbols GLUD2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 319-558 of human GLUD2 (NP_0362162).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2747
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.