Learn More
Invitrogen™ GluD2 Polyclonal Antibody

Description
GluD2 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Specifications
Specifications
| Antigen | GluD2 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.48 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | GLUD2 |
| Gene Accession No. | P49448 |
| Gene Alias | B230104L07Rik; cpr; creeper; GDH 2; GDH2; GluD2; GLUDP1; GluR delta2; gluR delta-2 subunit; GLURD2; GluRdelta2; glutamate dehydrogenase 2; glutamate dehydrogenase 2, mitochondrial; glutamate dehydrogenase pseudogene 1; glutamate ionotropic receptor delta type subunit 2; glutamate receptor delta-2 subunit; glutamate receptor ionotropic, delta-2; glutamate receptor, ionotropic, delta 2; Grid2; ho; hotfoot; Lc; Lc>J>; lurcher; minisatellite 10ac detected by probe MMS10; MMS10-AC; Ms10ac; nmf408; SCAR18; testicular secretory protein Li 14; tpr |
| Gene Symbols | GLUD2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.