Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GluR1, p Ser845 Antibody (SR1027), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321554100UL
Description
GluR1 Monoclonal antibody specifically detects GluR1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
GluR1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
SR1027 | |
Western Blot 1:500-1:1000 | |
AMPA 1, AMPA-selective glutamate receptor 1, GluA1, GLUH1, GluR-1, GLUR1gluR-A, GluR-A, GLURA, GluR-K1, glutamate receptor 1, Glutamate receptor ionotropic, AMPA 1, glutamate receptor, ionotropic, AMPA 1, HBGR1, MGC133252 | |
A synthesized peptide derived from human phospho-GluR1 [Ser845] (Uniprot #: P42261) | |
100 μg | |
Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Vision | |
2890 | |
Store at -20 degrees C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction