Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GluR1 Recombinant Rabbit Monoclonal Antibody (4H62L3)

Catalog No. 703312
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-331-2 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-331-2 Supplier Invitrogen™ Supplier No. 703312
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This antibody is predicted to react with Monkey and Bovine.

GluR1 (glutamate receptor 1) is an ionotropic glutamate receptor. L-glutamate acts as an excitatory neurotransmitter at many synapses in the central nervous system. Binding of the excitatory neurotransmitter, L-glutamate, induces a conformation change that leads to the opening of the cation channel and converts the chemical signal to an electrical impulse. Then, the receptor desensitizes rapidly and enters a transiet inactive state characterized by the presence of a bound agonist. In the presence of CACNG4, CACNG7, or CACNG8 GluR1 shows resensitization which is characterized by a delayed accumulation of current flux upon continued application of glutamate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GluR1
Applications Western Blot
Classification Recombinant Monoclonal
Clone 4H62L3
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene GRIA1
Gene Accession No. P19490, P23818, P42261
Gene Alias 2900051M01Rik; AI853806; AMPA 1; AMPA glutamate receptor; AMPA receptor GluR1/A; AMPA-selective glutamate receptor 1; Glr1; Glr-1; GluA1; GLUH 1; GLUH1; GluR K1; GLUR1; Glur-1; GluR1 flip; GluR1 flop; GLURA; GluR-A; GluRK1; gluR-K1; glutamate ionotropic receptor AMPA type subunit 1; glutamate receptor; glutamate receptor 1; glutamate receptor A; glutamate receptor ionotropic, AMPA 1; glutamate receptor subunit GluR1; glutamate receptor, ionotropic, AMPA 1; glutamate receptor, ionotropic, AMPA1 (alpha 1); GPRC1A; Gria 1; Gria1; GRM1A; HBGR1; HGBR1; HIPA; HIPA1; MGC13325; MGC133252; NMDA1; NMDAR1; NR1; OTTHUMP00000160643; OTTHUMP00000165781; OTTHUMP00000224241; OTTHUMP00000224242; OTTHUMP00000224243; RP11-350O14.1
Gene Symbols GRIA1
Host Species Rabbit
Immunogen Protein corresponding to human GluR1 [aa121-aa300].
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14799, 2890, 50592
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.