Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GluR2/3 Antibody (SR2311), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | GluR2/3 |
---|---|
Clone | SR2311 |
Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1:100 |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Description
GluR2/3 Monoclonal antibody specifically detects GluR2/3 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
GluR2/3 | |
Western Blot 1:500-1:2000, Immunoprecipitation 1:100 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
AMPA 2, glutamate receptor, ionotropic, AMPA 2 | |
A synthesized peptide derived from human GluR2/3 (Uniprot #: P42262, P42263) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR2311 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Tumor Suppressors, Vision | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
2891 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title