Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GluR2 Antibody (4C7), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
| Antigen | GluR2 |
|---|---|
| Clone | 4C7 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
Description
GluR2 Monoclonal antibody specifically detects GluR2 in Human samples. It is validated for ELISASpecifications
| GluR2 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| AMPA 2, glutamate receptor, ionotropic, AMPA 2 | |
| A synthesized peptide derived from Human GluR2 [UniProt P42262] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 4C7 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Tumor Suppressors, Vision | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 2891 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title