Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GluR2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB300301
Description
GluR2 Polyclonal specifically detects GluR2 in Rat samples. It is validated for Western Blot.Specifications
GluR2 | |
Polyclonal | |
Western Blot 1:1000 | |
P19491 | |
GRIA2 | |
Synthetic peptide corresponding to amino acid residues specific to GluR2 conjugated to KLH. Accession # P19491 | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Specific for the ∼100k GluR2 protein. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative | |
AMPA 2, glutamate receptor, ionotropic, AMPA 2 | |
Rabbit | |
100 kDa | |
0.1 mL | |
Apoptosis, Cancer, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Tumor Suppressors, Vision | |
2891 | |
Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction