Learn More
GluR2 (extracellular) Polyclonal Antibody, Invitrogen™
Guinea Pig Polyclonal Antibody
Supplier: Thermo Scientific PA5111823
Description
Reconstitute antibody with 50 μL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min). Peptide sequence is: NVGNINNDKK DETYR(C) Homology: Mouse amino acid residues identical; human - 14/15 amino acid residues identical.
The ion channels activated by glutamate are typically divided into two classes. Those that are sensitive to N-methyl-D-aspartate (NMDA) are designated NMDA receptors (NMDAR) while those activated by alpha-amino-3-hydroxy-5 -methyl-4-isoxalone propionic acid (AMPA) are known as AMPA receptors (AMPAR). The AMPAR are comprised of four distinct Glutamate Receptor Subunits designated (GluR1-4) and they play key roles in virtually all excitatory neurotransmission in the brain. The GluR2 subunit is thought to play a key role in forms of synaptic plasticity such as LTD.
Specifications
GluR2 (extracellular) | |
Polyclonal | |
Unconjugated | |
Gria2 | |
AMPA-selective glutamate receptor 2; GluA2; GLUR2; GluR-2; GLURB; GluR-B; GluR-K2; Glutamate receptor 2; Glutamate receptor ionotropic, AMPA 2; GRIA2; HBGR2; OTTHUMP00000165324; OTTHUMP00000220942; OTTHUMP00000220943 | |
Guinea Pig | |
Antigen affinity chromatography | |
RUO | |
14800, 29627 | |
-20°C | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunomicroscopy, Western Blot | |
0.8 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
P19491, P23819 | |
Gria2 | |
Peptide corresponding to amino acid residues 179-193 of rat GluR2 (extracellular, N-terminus) | |
50 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.