Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GluR2/GluR3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA14660
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA14660 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA14660 Supplier Invitrogen™ Supplier No. PA14660
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with chicken, human, mouse and zebrafish based on 100% sequence homology. This antibody is specific for the ~100 kDa GluR2/3 protein.

Glutamate receptors are the predominant excitatory neurotransmitter receptors in the mammalian brain and are activated in a variety of normal neurophysiologic processes. These receptors are heteromeric protein complexes composed of multiple subunits, arranged to form ligand-gated ion channels. The classification of glutamate receptors is based on their activation by different pharmacologic agonists. The subunit encoded by this gene belongs to a family of AMPA (alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate)-sensitive glutamate receptors, and is subject to RNA editing (AGA->GGA; R->G). Alternative splicing at this locus results in different isoforms, which may vary in their signal transduction properties.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GluR2/GluR3
Applications Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene Gria2
Gene Accession No. P19491, P19492
Gene Alias 2900064I19Rik; AMPA selective glutamate receptor; AMPA-selective glutamate receptor 2; AMPA-selective glutamate receptor 3; dJ1171F9.1; GluA2; GluA3; GLUR2; GluR-2; GLUR3; GluR-3; Gluralpha3; GLURB; gluR-B; GLURC; GLUR-C; GluR-K2; GluR-K3; glutamate ionotropic receptor AMPA type subunit 2; glutamate ionotropic receptor AMPA type subunit 3; glutamate receptor; glutamate receptor 2; Glutamate receptor 3; glutamate receptor B; glutamate receptor B flip isoform; glutamate receptor C; glutamate receptor channel alpha3 subunit; glutamate receptor ionotropic, AMPA 2; Glutamate receptor ionotropic, AMPA 3; glutamate receptor subunit 3; glutamate receptor, ionotrophic, AMPA 3; glutamate receptor, ionotropic, AMPA 2; glutamate receptor, ionotropic, AMPA 3; glutamate receptor, ionotropic, AMPA2 (alpha 2); glutamate receptor, ionotropic, AMPA3 (alpha 3); GPRC1C; GRIA2; GRIA3; GRM3; HBGR2; Kiaa4184; MGR3; MRX94
Gene Symbols Gria2, Gria3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the C-terminal region of rat GRIA2/3 conjugated to KLH.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29627, 29628
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.