Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GluR2/GluR3 Recombinant Rabbit Monoclonal Antibody (4C3)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA549790
Description
GluR2/GluR3 Recombinant Monoclonal Antibody for ICC/IF, ELISA
Glutamate receptors are the predominant excitatory neurotransmitter receptors in the mammalian brain and are activated in a variety of normal neurophysiologic processes. These receptors are heteromeric protein complexes composed of multiple subunits, arranged to form ligand-gated ion channels. The classification of glutamate receptors is based on their activation by different pharmacologic agonists. The subunit encoded by this gene belongs to a family of AMPA (alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate)-sensitive glutamate receptors, and is subject to RNA editing (AGA->GGA; R->G). Alternative splicing at this locus results in different isoforms, which may vary in their signal transduction properties.
Specifications
GluR2/GluR3 | |
Recombinant Monoclonal | |
1.5 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P42262, P42263 | |
Gria2, Gria3 | |
A synthesized peptide derived from human GRIA2/GRIA3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Immunocytochemistry | |
4C3 | |
Unconjugated | |
Gria2 | |
2900064I19Rik; AMPA selective glutamate receptor; AMPA-selective glutamate receptor 2; AMPA-selective glutamate receptor 3; dJ1171F9.1; GluA2; GluA3; GLUR2; GluR-2; GLUR3; GluR-3; Gluralpha3; GLURB; gluR-B; GLURC; GLUR-C; GluR-K2; GluR-K3; glutamate ionotropic receptor AMPA type subunit 2; glutamate ionotropic receptor AMPA type subunit 3; glutamate receptor; glutamate receptor 2; Glutamate receptor 3; glutamate receptor B; glutamate receptor B flip isoform; glutamate receptor C; glutamate receptor channel alpha3 subunit; glutamate receptor ionotropic, AMPA 2; Glutamate receptor ionotropic, AMPA 3; glutamate receptor subunit 3; glutamate receptor, ionotrophic, AMPA 3; glutamate receptor, ionotropic, AMPA 2; glutamate receptor, ionotropic, AMPA 3; glutamate receptor, ionotropic, AMPA2 (alpha 2); glutamate receptor, ionotropic, AMPA3 (alpha 3); GPRC1C; GRIA2; GRIA3; GRM3; HBGR2; Kiaa4184; MGR3; MRX94 | |
Rabbit | |
Affinity chromatography | |
RUO | |
2891, 2892 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction