Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ GluR2, Mouse, Unlabeled, Clone: 14C12.2,

Catalog No. MABN1189 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABN1189 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1189 Supplier MilliporeSigma™ Supplier No. MABN1189
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Glutamate receptor 2 (UniProt P19491; also known as AMPA-selective glutamate receptor 2, GluA2, GluR-2, GluR-B, GluR-K2, GluR2, Glutamate receptor ionotropic, AMPA 2) is encoded by the Gria2 (also known as Glur2) gene (Gene ID 29627) in rat species. Glutamate receptors are synaptic receptors located primarily on the membranes of neuronal cells. These receptors play important roles in neural communication, memory formation, learning, and regulation by mediating glutamate-dependent postsynaptic excitation. Glutamate receptors are divided into two types, ionotropic glutamate receptors (iGluRs) and metabotropic glutamate receptors (mGluRs). iGluRs form ion channel pores upon glutamate binding, while mGluRs indirectly activate ion channels on the plasma membrane via G protein-coupled signaling cascade. Although all iGluRs and mGluRs bind glutamate, they do exhibit different binding activities toward chemical ligands, such as NMDA, kainate, AMPA, L-AP4, ACPD, L-QA, and such distinciton is used to divide these receptors further into subtypes. Glutamate receptor 2 is an AMPA type iGluR, GluR-1 is initially produced with an N-terminal signal peptide sequence (a.a. 1-24), the removal of which yields the mature receptor with 4 transmembrane helices (a.a. 544-564, 592-610, 617-637, 813-833), having a large N-terminal extracellular domain (a.a. 24-543), followed by two intracellular loops (a.a. 565-591, 611-616), one extracellular loop (a.a. 638-812) and a C-terminal cytoplasmic tail (a.a. 834-883).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GluR2
Applications Western Blot
Classification Monoclonal
Clone 14C12.2
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG2a in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene Gria2, Glur2
Gene Accession No. P19491
Host Species Mouse
Immunogen GST-tagged recombinant rat GluR2 N-terminal extracellular domain fragment.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_001077280
Test Specificity Clone 14C12.2 targets an epitope within the N-terminal extracellular domain present in all spliced isoforms of human, mouse, and rat GluR2 reported by UniProt (P42262, P23819, P19491).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.