Learn More
Invitrogen™ GluR2 Polyclonal Antibody

Description
Immunogen sequence: NSIQIGGLFP RGADQEYSAF RVGMVQFSTS EFRLTPHIDN LEVANSFAVT NAFCSQFSRG VYAIFGFYDK KSVNTITSFC GTLHVSFITP SFPTDGTHPF VIQMRPDLKG ALLSLIEYYQ WDKFAYLYDS DRGLSTLQAV LDSAAEKKWQ VTAINVGNIN NDKKDEMYRS LFQDLELKKE RRVILDCERD KVNDIVDQVI TIGKHV; Positive Samples: SH-SY5Y, HL-60, Mouse liver, Mouse brain, Mouse pancreas, Rat brain; Cellular Location: Cell junction, Cell membrane, Endoplasmic reticulum membrane, Multi-pass membrane protein, postsynaptic cell membrane, synapse.
Specifications
Specifications
| Antigen | GluR2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.52 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Gria2 |
| Gene Accession No. | P23819 |
| Gene Alias | AMPA selective glutamate receptor; AMPA-selective glutamate receptor 2; GluA2; Glur2; GluR-2; GLURB; GluR-B; GluR-K2; glutamate ionotropic receptor AMPA type subunit 2; glutamate receptor; glutamate receptor 2; glutamate receptor B; glutamate receptor B flip isoform; glutamate receptor ionotropic, AMPA 2; glutamate receptor, ionotropic, AMPA 2; glutamate receptor, ionotropic, AMPA2 (alpha 2); GRIA2; HBGR2; OTTHUMP00000165324; OTTHUMP00000220942; OTTHUMP00000220943 |
| Gene Symbols | Gria2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.