Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GluR3 Monoclonal Antibody (1D2)
GREENER_CHOICE

Catalog No. PIMA515869
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515869 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515869 Supplier Invitrogen™ Supplier No. MA515869
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15869 targets GRIA3 in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15869 immunogen is purified recombinant fragment of human GRIA3 expressed in E. Coli. . MA5-15869 detects GRIA3 which has a predicted molecular weight of approximately 101kDa.

Glutamate receptors are the predominant excitatory neurotransmitter receptors in the mammalian brain and are activated in a variety of normal neurophysiologic processes. These receptors are heteromeric protein complexes composed of multiple subunits, arranged to form ligand-gated ion channels. The classification of glutamate receptors is based on their activation by different pharmacologic agonists. The subunit encoded by this gene belongs to a family of AMPA (alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate)-sensitive glutamate receptors, and is subject to RNA editing (AGA->GGA; R->G). Alternative splicing at this locus results in different isoforms, which may vary in their signal transduction properties.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GluR3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1D2
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.03% sodium azide
Gene GRIA3
Gene Accession No. P42263
Gene Alias 2900064I19Rik; AMPA-selective glutamate receptor 3; dJ1171F9.1; GluA3; GLUR3; Glur-3; Gluralpha3; GLURC; GLUR-C; GluR-K3; glutamate ionotropic receptor AMPA type subunit 3; glutamate receptor; glutamate receptor 3; glutamate receptor C; glutamate receptor channel alpha3 subunit; Glutamate receptor ionotropic, AMPA 3; glutamate receptor subunit 3; glutamate receptor, ionotrophic, AMPA 3; glutamate receptor, ionotropic, AMPA 3; glutamate receptor, ionotropic, AMPA3 (alpha 3); GRIA3; Kiaa4184; LOW QUALITY PROTEIN: glutamate receptor 3; MRX94; OTTHUMP00000024262; RP11-349N19.3
Gene Symbols GRIA3
Host Species Mouse
Immunogen Purified recombinant fragment of human GRIA3 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2892
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.