Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GluR4 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518931
Description
This antibody is predicted to react with bovine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
Glutamate receptors mediate most excitatory neurotransmission in the brain and play an important role in neural plasticity, neural development and neurodegeneration. Ionotropic glutamate receptors are categorized into NMDA receptors and kainate/AMPA receptors, both of which contain glutamate-gated, cation-specific ion channels. Kainate/AMPA receptors are co-localized with NMDA receptors in many synapses and consist of seven structurally related subunits designated GluR-1 to -7. The kainate/AMPA receptors are primarily responsible for the fast excitatory neuro-transmission by glutamate, whereas the NMDA receptors are functionally characterized by a slow kinetic and a high permeability for Ca2+ ions.
Specifications
GluR4 | |
Polyclonal | |
Unconjugated | |
Gria4 | |
AMPA-selective glutamate receptor 4; GluA4; GLUR4; Glur-4; GLUR4C; Gluralpha4; GLURD; GluR-D; glutamate ionotropic receptor AMPA type subunit 4; glutamate receptor; glutamate receptor 4; glutamate receptor channel alpha 4; glutamate receptor ionotropic, AMPA 4; glutamate receptor, ionotrophic, AMPA 4; glutamate receptor, ionotropic, AMPA 4; glutamate receptor, ionotropic, AMPA4 (alpha 4); Gria4; spike wave 1; spkw1 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
2893 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P48058 | |
Gria4 | |
Synthetic peptide sequence (KKLDQREYPGSETP) corresponding to the internal amino acids of GRIA4 (aa 280-294). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction