Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GluR4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116603
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116603 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116603 Supplier Invitrogen™ Supplier No. PA5116603
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: SH-SY5Y, B cells, Mouse brain, Mouse liver, Mouse kidney, Mouse spinal cord, Rat brain Immunogen sequence: HGGANVTGFQ LVDFNTPMVI KLMDRWKKLD QREYPGSETP PKYTSALTYD GVLVMAETFR SLRRQKIDIS RRGNAGDCLA NPAAPWGQGI DMERTLKQVR IQGLTGNVQF DHYGRRVNYT MDVFELKSTG PRKVGYWNDM DKLVLIQDVP TLGNDTAAIE NRTVVVTTIM PLMKNPILRN.

Glutamate receptors mediate most excitatory neurotransmission in the brain and play an important role in neural plasticity, neural development and neurodegeneration. Ionotropic glutamate receptors are categorized into NMDA receptors and kainate/AMPA receptors, both of which contain glutamate-gated, cation-specific ion channels. Kainate/AMPA receptors are co-localized with NMDA receptors in many synapses and consist of seven structurally related subunits designated GluR-1 to -7. The kainate/AMPA receptors are primarily responsible for the fast excitatory neuro-transmission by glutamate, whereas the NMDA receptors are functionally characterized by a slow kinetic and a high permeability for Ca2+ ions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GluR4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.92 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Gria4
Gene Accession No. P19493, P48058, Q9Z2W8
Gene Alias AMPA-selective glutamate receptor 4; GluA4; GLUR4; Glur-4; GLUR4C; Gluralpha4; GLURD; GluR-D; glutamate ionotropic receptor AMPA type subunit 4; glutamate receptor; glutamate receptor 4; glutamate receptor channel alpha 4; glutamate receptor ionotropic, AMPA 4; glutamate receptor, ionotrophic, AMPA 4; glutamate receptor, ionotropic, AMPA 4; glutamate receptor, ionotropic, AMPA4 (alpha 4); Gria4; spike wave 1; spkw1
Gene Symbols Gria4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 254-433 of human GluR4/GluA4/GluR4/GluA4/GRIA4 (NP_0010707121).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14802, 2893, 29629
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.