Learn More
GluR4 Polyclonal Antibody, Abbomax
Rabbit Polyclonal Antibody
Supplier: Abbomax Inc 5002904

Description
Positive control: Brain tissue Cellular location: Cytoplasmic, membrane.
GluR4 Polyclonal antibody specifically detects GluR4 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western BlotSpecifications
GluR4 | |
Polyclonal | |
Unconjugated | |
Gria4 | |
AMPA-selective glutamate receptor 4; GluA4; GLUR4; Glur-4; GLUR4C; Gluralpha4; GLURD; GluR-D; glutamate ionotropic receptor AMPA type subunit 4; glutamate receptor; glutamate receptor 4; glutamate receptor channel alpha 4; glutamate receptor ionotropic, AMPA 4; glutamate receptor, ionotrophic, AMPA 4; glutamate receptor, ionotropic, AMPA 4; glutamate receptor, ionotropic, AMPA4 (alpha 4); Gria4; spike wave 1; spkw1 | |
A synthetic peptide corresponding to the C-terminus of Glutamate R4 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.5 mg/mL | |
PBS with proprietary stabilizer and 0.01% sodium azide | |
P19493, P48058, Q9Z2W8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14802, 2893, 29629 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.