Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GluR6/7 Polyclonal Antibody, Abbomax

Catalog No. 502203677
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3677 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3677 Supplier Abbomax Inc Supplier No. 5002934
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Brain Cellular location: Cytoplasmic, Membrane.

GluR6/7 Polyclonal antibody specifically detects GluR6/7 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot

Specifications

Antigen GluR6/7
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene Grik2
Gene Accession No. B1AS29, P39087, P42260, P42264, Q13002, Q13003
Gene Alias bA487F5.1; dJ1090M5.1 (glutamate receptor, ionotropic, kainate 3 (GLUR7)); EAA4; EAA5; excitatory amino acid receptor 4; Excitatory amino acid receptor 5; GLR6; GLR7; GluK2; GluK3; GLUK6; GLUR6; GluR-6; Glur7; gluR-7; GluR7a; glutamate ionotropic receptor kainate type subunit 2; glutamate ionotropic receptor kainate type subunit 3; glutamate receptor; glutamate receptor 6; Glutamate receptor 7; glutamate receptor form A; glutamate receptor form B; glutamate receptor form C; glutamate receptor form D; glutamate receptor form E; glutamate receptor ionotropic, kainate 2; Glutamate receptor ionotropic, kainate 3; glutamate receptor subunit kainate subtype; glutamate receptor, ionotropic, kainate 2; glutamate receptor, ionotropic, kainate 3; GRIK2; GRIK3; MRT6
Host Species Rabbit
Immunogen A synthetic peptide derived from C-terminus of Glutamate receptor 6/7 Protein
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14806, 14807, 2898, 2899, 298521, 54257
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.