Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GluR6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541081
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541081 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541081 Supplier Invitrogen™ Supplier No. PA541081
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PDFSSLSRAI LDLVQFFKWK TVTVVYDDST GLIRLQELIK APSRYNLRLK Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

Glutamate receptors are the primary excitatory neurotransmitter receptors in the mammalian brain and are activated during various normal neurophysiological processes. The kainate family of glutamate receptors, which are composed of four subunits and function as ligand-activated ion channels, includes this gene product. The subunit encoded by this gene is edited at multiple sites within the first and second transmembrane domains, which is thought to alter the structure and function of the receptor complex. Furthermore, different isoforms of this gene have been described due to alternatively spliced transcript variants. Mutations in this gene have been linked to autosomal recessive cognitive disability. GRIK2-associated diseases include Neurodevelopmental Disorder with Impaired Language and Ataxia and with or without Seizures and Intellectual Developmental Disorder, Autosomal Recessive 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GluR6
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GRIK2
Gene Accession No. P39087, Q13002
Gene Alias AW124492; bA487F5.1; EAA4; Excitatory amino acid receptor 4; GLR 6; GLR6; GluK2; GLUK6; GLUR 6; gluR beta-2; Glur6; Glur-6; Glurbeta2; glutamate ionotropic receptor kainate type subunit 2; glutamate receptor; glutamate receptor 6; Glutamate receptor beta-2; glutamate receptor form A; glutamate receptor form B; glutamate receptor form C; glutamate receptor form D; glutamate receptor form E; glutamate receptor ionotropic, kainate 2; glutamate receptor, ionotropic kainate 2; glutamate receptor, ionotropic, kainate 2; glutamate receptor, ionotropic, kainate 2 (beta 2); glutamate receptor, ionotropic, kainate 5; GRIK 2; Grik2; GRIK2 protein; grik2alpha; grik2l; MRT6
Gene Symbols GRIK2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human GRIK2 (aa 151-200).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14806, 2898
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.