Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GluR7/GRIK3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | GluR7/GRIK3 |
---|---|
Concentration | LYOPH |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
GluR7/GRIK3 Polyclonal specifically detects GluR7/GRIK3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
GluR7/GRIK3 | |
Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
Unconjugated | |
Rabbit | |
Neuronal Cell Markers, Neurotransmission | |
2899 | |
IgG | |
Unpurified | |
Specific for GRIK3. |
LYOPH | |
Polyclonal | |
Antisera | |
RUO | |
Rat, Mouse, Human | |
GRIK3 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title