Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GluR7/GRIK3 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176387DL594
Description
GluR7/GRIK3 Polyclonal antibody specifically detects GluR7/GRIK3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISASpecifications
GluR7/GRIK3 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, ELISA | |
DyLight 594 | |
EAA5, Excitatory amino acid receptor 5, GLR7, gluR-7, GluR7, GluR7a, GLUR7dJ1090M5.1 (glutamate receptor, ionotropic, kainate 3 (GLUR7)), Glutamate receptor 7, glutamate receptor, ionotropic kainate 3, glutamate receptor, ionotropic, kainate 3 | |
Antibody was raised against a 13 amino acid synthetic peptide near the amino terminus of the human Grik3. The immunogen is located within amino acids 30 - 80 of Grik3. Amino Acid Squence: EYADGPNAQVMNA | |
0.1 mL | |
Neuronal Cell Markers, Neurotransmission | |
2899 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction