Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GluR7 Monoclonal Antibody (2B3D1)
GREENER_CHOICE

Catalog No. PIMA531743
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531743 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531743 Supplier Invitrogen™ Supplier No. MA531743
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31743 has been tested in indirect ELISA.

Glutamate receptors are the predominant excitatory neurotransmitter receptors in the mammalian brain and are activated in a variety of normal neurophysiologic processes. This gene product belongs to the kainate family of glutamate receptors, which are composed of four subunits and function as ligand-activated ion channels. It is not certain if the subunit encoded by this gene is subject to RNA editing as the other 2 family members (GRIK1 and GRIK2). A Ser310Ala polymorphism has been associated with schizophrenia, and there are conflicting reports of its association with the pathogenesis of delirium tremens in alcoholics.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GluR7
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 2B3D1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene GRIK3
Gene Accession No. Q13003
Gene Alias 9630027E11; dJ1090M5.1 (glutamate receptor, ionotropic, kainate 3 (GLUR7)); EAA5; Excitatory amino acid receptor 5; GLR7; GluK3; GLUR7; Glur-7; GluR7-3; GluR7a; glutamate ionotropic receptor kainate type subunit 3; glutamate receptor; glutamate receptor 7; Glutamate receptor ionotropic, kainate 3; glutamate receptor subunit kainate subtype; glutamate receptor, ionotropic, kainate 3; Grik3
Gene Symbols GRIK3
Host Species Mouse
Immunogen Purified recombinant fragment of human GRIK3 (AA: extra 32-173) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2899
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.