Learn More
Invitrogen™ GluR7 Polyclonal Antibody

Description
A suggested positive control is human brain tissue lysate. PA5-20450 can be used with blocking peptide PEP-0567.
Specifications
Specifications
| Antigen | GluR7 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | GRIK3 |
| Gene Accession No. | B1AS29, P42264, Q13003 |
| Gene Alias | 9630027E11; dJ1090M5.1 (glutamate receptor, ionotropic, kainate 3 (GLUR7)); EAA5; Excitatory amino acid receptor 5; GLR7; GluK3; GLUR7; Glur-7; GluR7-3; GluR7a; glutamate ionotropic receptor kainate type subunit 3; glutamate receptor; glutamate receptor 7; Glutamate receptor ionotropic, kainate 3; glutamate receptor subunit kainate subtype; glutamate receptor, ionotropic, kainate 3; Grik3 |
| Gene Symbols | GRIK3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.