Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ GLUT-2, Rabbit, Unlabeled, Polyclonal,

Catalog No. 071402IMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
071402IMI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 071402IMI Supplier MilliporeSigma™ Supplier No. 071402I
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Solute carrier family 2, facilitated glucose transporter member 2 (UniProt P14246; also known as Glucose transporter type 2, liver, GLUT-2) is encoded by the Slc2a2 (also known as Glut-2, Glut2) gene (Gene ID 20526) in murine species. GLUTs (Glucose transporters) are a family of integral membrane glycoproteins that facilitative glucose uptake by cells. Molecular cloning of glucose transporters have identified a family of closely related genes that encode at least 7 proteins exhibiting high degree of homology (45% to 65%). GLUT2 is a facilitative glucose transporter that mediates the bidirectional transfer of glucose across the hepatocyte membrane. It is present at high levels in the plasma membrane of pancreatic beta cells where it initiates the first step in glucose-stimulated insulin secretion. Due to its low affinity and high capacity, GLUT2 transports dietary sugars, glucose, fructose and galactose in a large range of physiological concentrations, displaying large bidirectional fluxes in and out the cells. It also participates in the transport of glucose in small intestine and kidney along with the Na+/glucose cotransporter. Hepatic GLUT2 expression is upregulated in a dose-dependent manner according to glucose concentration in primary hepatocytes and hepatocyte cell lines. Also, hepatic GLUT2 expression is reduced during starvation and returns to normal levels after refeeding. In diabetic animals, hepatic GLUT2 expression is increased, but is corrected to normal levels when hyperglycemic conditions are corrected by exogenous insulin. Ref.: Burkhardt, BR., et al. (2005). FEBS Lett. 579, 5759 5764.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLUT-2
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene Slc2a2, Glut-2, Glut2
Gene Accession No. P14246
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to a 16-a.a. sequence within the first extracellular loop of mouse GLUT-2.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_112474
Test Specificity This polyclonal antibody targets a region within the first extracellular loop of human, mouse, and rat GLUT-2.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.