Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glut1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 23 publications
$317.50 - $706.50
Specifications
Antigen | Glut1 |
---|---|
Dilution | Western Blot 1:5000, ELISA 1:12000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB300666SS
![]() |
Novus Biologicals
NB300666SS |
0.025 mg |
Each for $317.50
|
|
|||||
NB300666
![]() |
Novus Biologicals
NB300666 |
0.1 mg |
Each for $706.50
|
|
|||||
Description
Glut1 Polyclonal specifically detects Glut1 in Human, Mouse, Rat, Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
Glut1 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse, Rat, Zebrafish | |
P11166 | |
6513 | |
This Glut1 antibody is made against a synthetic peptide conjugated to KLH, corresponding to amino acids 478-492 of Human Glucose Transporter GLUT1. Peptide was covalently modified. Sequence: PEELFHPLGADSQV | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. | |
54.1 kDa |
Western Blot 1:5000, ELISA 1:12000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Cancer, Cardiovascular Biology, Cellular Markers, Diabetes Research, Lipid and Metabolism, Membrane Trafficking and Chaperones, Neuroscience, Plasma Membrane Markers, Signal Transduction | |
Tris/Glycine buffer (pH 7.4 - 7.8), HEPES, 0.5% BSA, 30% glycerol with 0.02% Sodium Azide | |
Choreoathetosis/Spasticity, Episodic (Paroxysmal Choreoathetosis), CSE, DYT17, DYT18, DYT9, EIG12, Glucose transporter type 1, erythrocyte/brain, GLUT-1, GLUT1DS, HepG2 glucose transporter, HTLVR, Human T-Cell Leukemia Virus (I and II) Receptor, MGC141895, MGC141896, PED, solute carrier family 2 (facilitated glucose transporter), member 1, Solute Carrier Family 2 Member 1, solute carrier family 2, facilitated glucose transporter member 1 | |
SLC2A1 | |
IgG | |
Affinity Purified | |
Glucose Transporter GLUT1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title