Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glut1 Mouse anti-Human, Clone: rGLUT1/2476, Novus Biologicals™
SDP

Catalog No. p-7234827 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB123242 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB123242 Supplier Novus Biologicals Supplier No. NBP30788420UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Glut1 Monoclonal specifically detects Glut1 in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin.

Specifications

Antigen Glut1
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rGLUT1/2476
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution ELISA 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml
Formulation 10 mM PBS with 0.05% BSA
Gene Alias Choreoathetosis/Spasticity, Episodic (Paroxysmal Choreoathetosis), CSE, DYT17, DYT18, DYT9, EIG12, Glucose transporter type 1, erythrocyte/brain, GLUT-1, GLUT1DS, HepG2 glucose transporter, HTLVR, Human T-Cell Leukemia Virus (I and II) Receptor, MGC141895, MGC141896, PED, solute carrier family 2 (facilitated glucose transporter), member 1, Solute Carrier Family 2 Member 1, solute carrier family 2, facilitated glucose transporter member 1
Host Species Mouse
Immunogen Recombinant fragment of human Glut1 protein (around aa 203-305) (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Cellular Markers, Diabetes Research, Lipid and Metabolism, Membrane Trafficking and Chaperones, Neuroscience, Plasma Membrane Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6513
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.