Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLUT1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA146152

Catalog No. PA146152


Description

Description

The target sequence has 100% sequence homology with primate, and 93% with rat, bovine, and rabbit.

GLUT1 Polyclonal antibody specifically detects GLUT1 in Human, Mouse, Rat samples. It is validated for ChIP Assay, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), In vitro Assay, Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GLUT1
Polyclonal
Unconjugated
SLC2A1
choreoathetosis/spasticity, episodic (paroxysmal choreoathetosis/spasticity); CSE; DYT17; DYT18; DYT9; EIG12; glucose transporter glut1; glucose transporter type 1, erythrocyte/brain; glucose transporter type I; GLUT; Glut1; Glut-1; GLUT1DS; GLUTB; GT1; GTG1; Gtg3; HepG2 glucose transporter; HTLVR; human T-cell leukemia virus (I and II) receptor; MGC141895; MGC141896; PED; RATGTG1; receptor for HTLV-1 and HTLV-2; SDCHCN; Slc2a1; solute carrier family 2 (facilitated glucose transporter), member 1; Solute carrier family 2 a 1 (facilitated glucose transporter) brain; solute carrier family 2 member 1; solute carrier family 2, facilitated glucose transporter member 1; solute carrier family 2, member 1
Rabbit
Antigen affinity chromatography
RUO
20525, 6513
-20°C or -80°C if preferred
Liquid
ChIP Assay, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), In vitro Assay, Western Blot
0.9 mg/mL
PBS with 0.02% sodium azide
P11166, P17809
SLC2A1
Human GLUT1 protein (between residues 1-100).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.