Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glut1 Rabbit anti-Human, Clone: GLUT1/3132R, Novus Biologicals™

Rabbit Monoclonal Antibody
$316.50
Specifications
| Antigen | Glut1 |
|---|---|
| Clone | GLUT1/3132R |
| Concentration | 0.2 mg/ml |
| Dilution | Flow Cytometry 1-2 ug/million cells, ELISA 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | Flow Cytometry, ELISA, Immunohistochemistry (Paraffin) |
Description
Glut1 Monoclonal specifically detects Glut1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry-Paraffin.Specifications
| Glut1 | |
| 0.2 mg/ml | |
| Flow Cytometry, ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cardiovascular Biology, Cellular Markers, Diabetes Research, Lipid and Metabolism, Membrane Trafficking and Chaperones, Neuroscience, Plasma Membrane Markers, Signal Transduction | |
| 10 mM PBS with 0.05% BSA | |
| 6513 | |
| IgG | |
| Protein A or G purified |
| GLUT1/3132R | |
| Flow Cytometry 1-2 ug/million cells, ELISA 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Choreoathetosis/Spasticity, Episodic (Paroxysmal Choreoathetosis), CSE, DYT17, DYT18, DYT9, EIG12, Glucose transporter type 1, erythrocyte/brain, GLUT-1, GLUT1DS, HepG2 glucose transporter, HTLVR, Human T-Cell Leukemia Virus (I and II) Receptor, MGC141895, MGC141896, PED, solute carrier family 2 (facilitated glucose transporter), member 1, Solute Carrier Family 2 Member 1, solute carrier family 2, facilitated glucose transporter member 1 | |
| Recombinant fragment of human Glut1 protein (around aa 203-305) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title