Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Glut1 Recombinant Protein Antigen
SDP

Catalog No. NBP254745X Shop All R&D Systems Products
Change view
Click to view available options
Protein Length:
GRTFDEIASGFRQGGASQSDKTPEELFHP
RNEENRAKSVLKKLRGTADVTHDLQE
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Protein Length
NBP254745X RNEENRAKSVLKKLRGTADVTHDLQE
NBP248609X GRTFDEIASGFRQGGASQSDKTPEELFHP
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP254745X Supplier Novus Biologicals™ Supplier No. NBP254745PEP
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human Glut1 The Glut1 Recombinant Protein Antigen is derived from E. coli. The Glut1 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.

Specifications

Protein Length RNEENRAKSVLKKLRGTADVTHDLQE
Purification Method >80% by SDS-PAGE and Coomassie blue staining
Common Name Glut1 Recombinant Protein Antigen
Content And Storage Store at −20°C. Avoid freeze-thaw cycles.
Formulation PBS (pH 7.4), 1 M Urea
For Use With (Application) Blocking/Neutralizing, Control
Gene Alias DYT17, DYT18, DYT18DYT17, DYT9, EIG12, Glucose transporter type 1, erythrocyte/brain, Glut1
Gene Symbol SLC2A1
Label Type Unlabeled
Product Type Recombinant Protein Antigen
Quantity 0.1mL
Regulatory Status RUO
Research Category Cancer, Cardiovascular Biology, Cellular Markers, Diabetes Research, Lipid and Metabolism, Membrane Trafficking and Chaperones, Neuroscience, Plasma Membrane Markers, Signal Transduction
Source E.coli
Specific Reactivity Human
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.