Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLUT10 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100490
Description
GLUT10 Polyclonal specifically detects GLUT10 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
GLUT10 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
SLC2A10 | |
A synthetic peptide containing residues 367-385 (ILSTAKKTKPHPRSGDPSA) of the human GLUT10 protein. | |
Affinity Purified | |
RUO | |
Primary | |
Human GLUT10 protein. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
1 mg/mL | |
Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
ATS, Glucose transporter type 10, GLUT-10, GLUT10MGC126706, solute carrier family 2 (facilitated glucose transporter), member 10, solute carrier family 2, facilitated glucose transporter member 10 | |
Rabbit | |
52 kDa | |
0.1 mg | |
Diabetes Research | |
81031 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction