Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLUT10 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298748PCP
Description
GLUT10 Polyclonal antibody specifically detects GLUT10 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
GLUT10 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
ATS, Glucose transporter type 10, GLUT-10, GLUT10MGC126706, solute carrier family 2 (facilitated glucose transporter), member 10, solute carrier family 2, facilitated glucose transporter member 10 | |
Produced in rabbits immunized with A synthetic peptide corresponding to the C-terminus of the human GLUT10. | |
0.1 mL | |
Diabetes Research | |
81031 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction