Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLUT10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583529
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583529 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583529 Supplier Invitrogen™ Supplier No. PA583529
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LPAGTDETAT HKDLIPLQGG EAPKLGPGRP RYSFLDLFRA RDNMRG Highest antigen sequence indentity to the following orthologs: Mouse - 63%, Rat - 61%.

Non-insulin-dependent diabetes mellitus (NIDDM) is a multifactoral disease with both environmental and genetics causes. Genome-wide screening procedures have identified several susceptibility loci for NIDDM within the human genome. A putative sugar transporter that has been localized to human chromosome 20q12-q13.1, one of the genomic loci associated with NIDDM. Because of the strong resemblance of this novel protein to members of the mammalian facilitative glucose transporter family (GLUT), the protein is known as GLUT10 (HGMW-approved gene symbol SLC2A10). Data suggests that GLUT10 an excellent candidate for a susceptibility gene involved in NIDDM.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLUT10
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC2A10
Gene Accession No. O95528
Gene Alias AA450473; ATS; glucose transporter type 10; Glut10; GLUT-10; GLUT11; MGC118830; MGC118833; Slc2a10; SLC2A11-a; SLC2A11-c; solute carrier family 2 (facilitated glucose transporter), member 10; solute carrier family 2 member 10; solute carrier family 2, facilitated glucose transporter member 10
Gene Symbols SLC2A10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC2A10. Recombinant protein control fragment (Product #RP-99462).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81031
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.