Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GLUT10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112069
Description
GLUT10 Polyclonal Antibody for Western Blot
Non-insulin-dependent diabetes mellitus (NIDDM) is a multifactoral disease with both environmental and genetics causes. Genome-wide screening procedures have identified several susceptibility loci for NIDDM within the human genome. A putative sugar transporter that has been localized to human chromosome 20q12-q13.1, one of the genomic loci associated with NIDDM. Because of the strong resemblance of this novel protein to members of the mammalian facilitative glucose transporter family (GLUT), the protein is known as GLUT10 (HGMW-approved gene symbol SLC2A10). Data suggests that GLUT10 an excellent candidate for a susceptibility gene involved in NIDDM.
Specifications
| GLUT10 | |
| Polyclonal | |
| PBS with no preservative | |
| O95528 | |
| SLC2A10 | |
| A synthetic peptide corresponding to the C-terminus of the Human SLC2A10. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| SLC2A10 | |
| AA450473; ATS; glucose transporter type 10; Glut10; GLUT-10; GLUT11; MGC118830; MGC118833; Slc2a10; SLC2A11-a; SLC2A11-c; solute carrier family 2 (facilitated glucose transporter), member 10; solute carrier family 2 member 10; solute carrier family 2, facilitated glucose transporter member 10 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 81031 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction