Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC2A12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580020
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: PC-12 Cell, A549 Cell.
Solute carrier family 2, facilitated glucose transporter member 12 is a protein that in humans is encoded by the SLC2A12 gene. SLC2A12 belongs to a family of transporters that catalyze the uptake of sugars through facilitated diffusion. This family of transporters show conservation of 12 transmembrane helices as well as functionally significant amino acid residues.
Specifications
SLC2A12 | |
Polyclonal | |
Unconjugated | |
SLC2A12 | |
facilitated glucose transporter; glucose transporter type 12; GLUT12; GLUT-12; GLUT8; SLC2A12; solute carrier family 2 (facilitated glucose transporter), member 12; solute carrier family 2 member 12; solute carrier family 2, facilitated glucose transporter member 12; solute carrier family 2, member 12 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
154091, 308028, 353169 | |
-20°C | |
Lyophilized |
Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
Q8BFW9, Q8TD20 | |
SLC2A12 | |
A synthetic peptide corresponding to a sequence in the middle region of human GLUT12 (256-272aa SLKDEYQYSFWDLFRSK). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction