Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLUT12 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984804
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4804 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4804 Supplier Bioss Supplier No. BS2540R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Solute carrier family 2, facilitated glucose transporter member 12 is a protein that in humans is encoded by the SLC2A12 gene. SLC2A12 belongs to a family of transporters that catalyze the uptake of sugars through facilitated diffusion. This family of transporters show conservation of 12 transmembrane helices as well as functionally significant amino acid residues.

Specifications

Antigen GLUT12
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SLC2A12
Gene Accession No. Q8TD20
Gene Alias BOS_9829; facilitated glucose transporter; Glucose transporter type 12; Glut12; GLUT-12; GLUT8; Slc2a12; solute carrier family 2 (facilitated glucose transporter), member 12; solute carrier family 2 member 12; Solute carrier family 2, facilitated glucose transporter member 12; solute carrier family 2, member 12
Gene Symbols SLC2A12
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GLUT12.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 154091
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.