Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glut2 Antibody (199017), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB1414APCCY7
Description
Glut2 Monoclonal antibody specifically detects Glut2 in Human samples. It is validated for Flow CytometrySpecifications
Glut2 | |
Monoclonal | |
APC-Cyanine7 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
199017 | |
Flow Cytometry | |
GLUT-2, GLUT2 Glucose transporter type 2, liver, SLC2A2, solute carrier family 2 (facilitated glucose transporter), member 2, solute carrier family 2, facilitated glucose transporter member 2 | |
NS0 mouse myeloma cell line transfected with human Glut2, Met1-Val524, Accession # P11168 | |
0.2 mL | |
Lipid and Metabolism, Stem Cell Signaling Pathway | |
6514 | |
Store at 4C in the dark. Do not freeze. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction