Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glut2 Antibody (205115), Janelia Fluor™ 646, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB1440J
This item is not returnable.
View return policy
Description
Glut2 Monoclonal antibody specifically detects Glut2 in Mouse samples. It is validated for Flow CytometrySpecifications
| Glut2 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| GLUT-2, GLUT2 Glucose transporter type 2, liver, SLC2A2, solute carrier family 2 (facilitated glucose transporter), member 2, solute carrier family 2, facilitated glucose transporter member 2 | |
| CHO Chinese hamster ovary cell line transfected with mouse Glut2, Met1-Val523, Accession # P14246 | |
| 0.2 mL | |
| Lipid and Metabolism, Stem Cell Signaling Pathway | |
| 6514 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry | |
| 205115 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction