Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLUT2 Monoclonal Antibody (199017)
GREENER_CHOICE

Catalog No. PIMA523806
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA523806 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA523806 Supplier Invitrogen™ Supplier No. MA523806
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody recognizes human Glut2 expression on human Glut2-transfected NS0 cells, but not on control transfectants. This antibody detects Glut2 on the surface of the human intestinal cell line Caco2 (2) in flow cytometry and on fixed cells in immunocytochemistry tests. Based on flow cytometric tests on transfected cells, this antibody has no cross-reactivity with human Glut1 or human Glut3. Reconstitute at 0.5 mg/mL in sterile PBS.

Glucose transporter 2 isoform is an integral plasma membrane glycoprotein of the liver, islet beta cells, intestine, and kidney epithelium. It mediates facilitated bidirectional glucose transport. Because of its low affinity for glucose, it has been suggested as a glucose sensor. Mutations in the gene that encode this protein cause Fanconi-Bickel syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLUT2
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 199017
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Slc2a2
Gene Accession No. P11168
Gene Alias AI266973; glucose transporter 2; glucose transporter type 2, liver; GLUT2; GLUT-2; GTT2; liver-type glucose transporter; Slc2a2; solute carrier family 2 (facilitated glucose transporter), member 2; solute carrier family 2 member 2; solute carrier family 2, facilitated glucose transporter member 2
Gene Symbols Slc2a2
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Glut2 Met1-Val524.
Purification Method Protein A/G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6514
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.