Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GLUT2 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142484
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:8,000.
Glucose transporter 2 isoform is an integral plasma membrane glycoprotein of the liver, islet beta cells, intestine, and kidney epithelium. It mediates facilitated bidirectional glucose transport. Because of its low affinity for glucose, it has been suggested as a glucose sensor. Mutations in the gene that encode this protein cause Fanconi-Bickel syndrome.
Specifications
| GLUT2 | |
| Polyclonal | |
| Unconjugated | |
| Slc2a2 | |
| AI266973; glucose transporter 2; glucose transporter type 2, liver; GLUT2; GLUT-2; GTT2; liver-type glucose transporter; Slc2a2; solute carrier family 2 (facilitated glucose transporter), member 2; solute carrier family 2 member 2; solute carrier family 2, facilitated glucose transporter member 2 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 6514 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P11168 | |
| Slc2a2 | |
| Peptide with sequence C-RKEREEASSEQKVS (aa 277-290). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction